Learn More
Cleatech CHEMICAL RESIST GLOVES 32"L,S8
SDP
Catalog No. NC2245917
Encompass_Preferred
Butadyl, Chemical Resistant, Anti-Static Dry Box Standard Gloves / 15-mil thick , 32" length with 8"-dia. cuff opening and 8 hand size)
This item is not returnable. View return policy
Catalog No. NC2245917 Supplier Cleatech Supplier No. A21GLB1508
Add to Cart
Add to Cart
Butadyl, Chemical Resistant, Anti-Static Dry Box Standard Gloves / 15-mil thick , 32" length with 8"-dia. cuff opening and 8 hand size)