Learn More
National Safety Apparel™ Chemical Resistant FR Lab Coats, No Pockets
SDP
Catalog No. 17000306 Shop All National Public Seating Corporation Products
Change view
Click to view available options
Size:
X-Small
Small
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-000-306 X-Small
17-000-307 Small
17-000-309 Large
17-000-310 X-Large
17-000-311 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 17-000-306 Supplier National Safety Apparel™ Supplier No. LABCRNPNBXS
Add to Cart
Add to Cart
Chemical Resistant Lab Coat made of flame-resistant CR/FR fabric for multi-hazard protection without weighing you down.
  • Fabric/Material 4.4 oz. CR/FR Aramid Blend FR Nomex knit wrists keep sleeves in place
  • Bottom side vents for increased mobility
  • Durable hi-temp plastic snaps for easy donning and doffing
  • Side pass-through allows access to pockets underneath the coat
  • Durable accessory loop for safety glasses, pens, badges, etc.
  • Highly breathable
  • Easy care and laundering
Specifications
Product Type Chemical Resistant Lab Coat
Size X-Small
Gender Unisex
Material Aramid Fiber
Color Navy
Closure Type Snaps
Cuff Style Knit
Disposable Reusable
Sterility Non-sterile
Resistance Chemicals, Flame
Style Knee-length
Certifications/Compliance ASTM F1506, NFPA 2112