Learn More
Grainger Honeywell Nitri-Knit Chemical-Resistant Gloves, Nitrile, 26 in., 10/XL, 6/Pk.
Catalog No. 502792494
Encompass_Preferred
These chemical-resistant gloves are constructed with a 100% nitrile formulation and feature an interlock knit liner for protection against a wide range of chemicals, including solvents, caustics, and animal fats. The extended sleeve design provides liquidproof coverage, while the rough finish ensures a secure grip in wet, oily, or dry conditions.
  • 100% nitrile formulation is free of latex proteins to prevent allergic reactions.
  • Interlock knit liner provides resistance to snags, cuts, abrasions, and punctures.
  • Thermal lining offers protection for temperatures ranging from -10°F to 300°F.
  • Curved hand shape reduces hand fatigue during extended use.
  • Anti-bacterial and anti-mildew treatment applied to the hand portion.
  • Complies with USDA and FDA regulations (21 CFR) for use in food processing.
  • Elastic cuff and shoulder elastic prevent slippage during use.
Catalog No. 50-279-2494 Supplier Grainger Supplier No. 1AHJ3
These chemical-resistant gloves are constructed with a 100% nitrile formulation and feature an interlock knit liner for protection against a wide range of chemicals, including solvents, caustics, and animal fats. The extended sleeve design provides liquidproof coverage, while the rough finish ensures a secure grip in wet, oily, or dry conditions.
  • 100% nitrile formulation is free of latex proteins to prevent allergic reactions.
  • Interlock knit liner provides resistance to snags, cuts, abrasions, and punctures.
  • Thermal lining offers protection for temperatures ranging from -10°F to 300°F.
  • Curved hand shape reduces hand fatigue during extended use.
  • Anti-bacterial and anti-mildew treatment applied to the hand portion.
  • Complies with USDA and FDA regulations (21 CFR) for use in food processing.
  • Elastic cuff and shoulder elastic prevent slippage during use.