Learn More
GA International Chemical-Resistant Labels for Resin-Embedding (Patented) 1" x 0.25" / 25.4mm x 6.4mm Rectangle White From -80°C to +149°C (-112°F to +300°F)
Catalog No. 5000570887
Encompass_Preferred
Hot resin and chemical-resistant labels for embedding in the resin capsule or mold, alongside the sample. The small size of the ResiTAG label can accommodate various sizes of embedding capsules commonly used in electron microscopy facilities. When printed with our recommended chemical-resistant resin ribbon (XAR-class, GA International), the ResiTAG label and printout withstand histological resins and oven incubation up to 150°C. In addition to the resin itself, the printout can also withstand ethanol and other alcohols, xylene, toluene, acetone, formalin, as well as other harsh chemicals, solvents, and histological stains.
Catalog No. 50-005-70887 Supplier GA International Supplier No. XST251C310WH
Hot resin and chemical-resistant labels for embedding in the resin capsule or mold, alongside the sample. The small size of the ResiTAG label can accommodate various sizes of embedding capsules commonly used in electron microscopy facilities. When printed with our recommended chemical-resistant resin ribbon (XAR-class, GA International), the ResiTAG label and printout withstand histological resins and oven incubation up to 150°C. In addition to the resin itself, the printout can also withstand ethanol and other alcohols, xylene, toluene, acetone, formalin, as well as other harsh chemicals, solvents, and histological stains.