Learn More
Lakeland Industries Chemical Shoe Cover
Catalog No. 502841120
Encompass_Preferred
Looking for next-level chemical protection? Start with Lakeland's ChemMax 2 product line. These 17" chemical boot covers are constructed with a Saranex 23P film and bound seams, making them an ideal option for chemical mixing and handling, environmental clean-up, and hazardous material remediation and response. Offered in assortments of 100 pair per case, C2B903P is available in sizes XL and 2X.
Catalog No. 50-284-1120 Supplier Lakeland Industries Supplier No. C2B903P2X
Looking for next-level chemical protection? Start with Lakeland's ChemMax 2 product line. These 17" chemical boot covers are constructed with a Saranex 23P film and bound seams, making them an ideal option for chemical mixing and handling, environmental clean-up, and hazardous material remediation and response. Offered in assortments of 100 pair per case, C2B903P is available in sizes XL and 2X.