Learn More
Lakeland Industries Chemical Shoe Cover
Catalog No. 502840449
Encompass_Preferred
Looking for next-level chemical protection? Start with Lakeland's ChemMax 2 product line. These 17" chemical boot covers are constructed with a Saranex 23P film and heat-sealed seams, making them an ideal option for chemical mixing and handling, environmental clean-up, and hazardous material remediation and response. Offered in cases of 12 pair per case, C2T740P is available in one size that should fit most wearers.
Catalog No. 50-284-0449 Supplier Lakeland Industries Supplier No. C2T740P
Looking for next-level chemical protection? Start with Lakeland's ChemMax 2 product line. These 17" chemical boot covers are constructed with a Saranex 23P film and heat-sealed seams, making them an ideal option for chemical mixing and handling, environmental clean-up, and hazardous material remediation and response. Offered in cases of 12 pair per case, C2T740P is available in one size that should fit most wearers.