Learn More
Neese Chemical Splash Coveralls
SDP
Catalog No. 19020324
Change view
Click to view available options
Size:
Medium
X-Large
Thickness:
1377.9 mil
35 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Thickness
19-020324 X-Large 1377.9 mil
19-168-351B Medium 35 mm
19-168-351D X-Large 35 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-020324 Supplier Neese Supplier No. 96001511GRNXL
Add to Cart
Add to Cart
This item is not returnable. View return policy
With an attached hood
  • Made of 35mm layered PVC/polyester/PVC
  • Resist many organic and inorganic acids, alcohols, and hydrocarbons
  • Inner sleeves and cuffs with welded seams for additional protection
  • Zipper- and snap-front with storm flap and welded seams
Specifications
Size X-Large
Material PVC, Polyester
Color Green
Neck Style Hooded
Closure Type Front Zipper with Snap and Storm Flap
Seam Type Welded
Description Attached hood
Thickness 1377.9 mil