Learn More
Pyramex™ Chemical Splash Goggle with Neoprene Strap
Catalog No. 19155317 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Lens Coating:
Anti-fog
Catalog No. Lens Coating
19-155-317 Anti-fog
1 options
This item is not returnable. View return policy
Catalog No. 19-155-317 Supplier Pyramex™ Supplier No. G304TN
Add to Cart
Add to Cart
This item is not returnable. View return policy
Clear H2X anti-fog
  • Body tinted to reduce glare
  • Special one-way vent allows air to enter while keeping liquids out
  • Top shelf chemical splash goggle features generously sized heavy duty body and lens
  • Scratch resistant polycarbonate lens provides 99% UVA/B/C protection
  • Specifications
    Lens Coating Anti-fog
    Lens Type Chemical-resistant
    Certifications/Compliance ANSI Z87.1
    Lens Tint Clear
    Product Type Clear H2X Anti-Fog with Neoprene Strap
    Frame Color Tinted
    Lens Material Polycarbonate
    Frame Material PVC