Learn More
MCR Safety Chemical Splash Goggles
Catalog No. 19037992 Shop All MCR Safety Products
Encompass_Preferred
Change view
Click to view available options
Product Type:
Safety Goggles
Catalog No. Product Type
19-037-992 Safety Goggles
1 options
This item is not returnable. View return policy
Catalog No. 19-037-992 Supplier MCR Safety Supplier No. 2230R
This item is not returnable. View return policy
These goggles provide a wide, unobstructed field of vision and can be worn with most half-mask respirators. Available in vented, perforated, chemical splash resistant and ventless styles
  • PVC body permits a comfortable fit over prescription eyewear
  • Options available to protect against impact and/or chemical splash
  • Rugged polycarbonate lens filters 99.9% of harmful U.V. rays
  • Exclusive Duramass™ scratch resistant coating
  • Goggle straps contain No Latex
California Prop 65 Warning: This product contains chemicals known to the State of California to cause cancer, birth defects or other reproductive harm.
Compliance
Meets ANSI Z87+, ANSI D3, and ANSI D4 standards.
Specifications
Certifications/Compliance ANSI Z87+, ANSI D3, ANSI D4
Lens Tint Clear
Product Type Safety Goggles
Ventilation Details Indirect
Material Polycarbon
Frame Material PVC