Learn More
MCR Safety Chemical Splash Goggles
Catalog No. 19065166 Shop All MCR Safety Products
Encompass_Preferred
Change view
Click to view available options
Product Type:
Chemical Splash Protection Goggles
Safety Goggles
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
19-065-166 Chemical Splash Protection Goggles
19-037-992 Safety Goggles
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-065-166 Supplier MCR Safety Supplier No. 2235R
Add to Cart
Add to Cart
This item is not returnable. View return policy
  • PVC body permits a comfortable fit over prescription eyewear
  • Options available to protect against impact and/or chemical splash
  • Rugged polycarbonate lens filters 99.9% of harmful U.V. rays
  • Exclusive Duramass™ scratch resistant coating
  • Goggle straps contain No Latex
California Prop 65 Warning: This product contains chemicals known to the State of California to cause cancer, birth defects or other reproductive harm.
Compliance
Meets ANSI Z87+, ANSI D3, and ANSI D4 standards.
Specifications
Certifications/Compliance ANSI Z87.1
Lens Coating Scratch-resistant
Lens Tint Clear
Product Type Chemical Splash Protection Goggles
Ventilation Details Indirect
Material Polycarbonate
Frame Material PVC