Learn More
Fisherbrand™ Chemical Splash and Impact Goggles
Catalog No. p-6837028 Shop All Fisherbrand Products
Change view
Click to view available options
Product Type:
Chemical Splash Protection Goggles
Impact Goggles
Ventilation Details:
Four indirect vents
Perforations on goggle top and sides provide direct ventilation
Certifications/Compliance:
ANSI/ISEA Z87.1-2020: Z87+ U6
ANSI/ISEA Z87.1-2020: Z87+ U6 D3
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Ventilation Details Certifications/Compliance
19-181-504 Chemical Splash Protection Goggles Four indirect vents ANSI/ISEA Z87.1-2020: Z87+ U6 D3
19-181-503 Impact Goggles Perforations on goggle top and sides provide direct ventilation ANSI/ISEA Z87.1-2020: Z87+ U6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-181-504
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Lens coated to minimize scratching and fogging.
Lens coated to minimize scratching and fogging
  • Built for comfortable protection with a soft, flexible PVC body that conforms to facial contours
  • Large frames fit over most prescription eyewear
  • Coated, clear polycarbonate lens resists impact, scratching, and fogging
  • Headstrap easily adjusts for a custom fit
  • Resist a wide range of chemicals and feature indirect ventilation to direct airflow over the lens
Order Info
NOTE: 19-181-503 is the IMPACT style goggles; 19-181-504 are the CHEMICAL SPLASH style goggles.
Compliance
Meets ANSI Z87.1 and ANSI Z80.3 standards.
Specifications
Certifications/Compliance ANSI/ISEA Z87.1-2020: Z87+ U6 D3
Lens Coating Anti-fog, Scratch-resistant
Shape Round
Lens Tint Clear
Product Type Chemical Splash Protection Goggles
Frame Color Green
Ventilation Details Four indirect vents
Material Polycarbonate
Lens Material Polycarbonate
Frame Material PVC