Learn More
Innovating Science™ pH Indicator Set Kit
GREENER_CHOICESCI_ED
Catalog No. S98668 Shop All Aldon Products
Change view
Click to view available options
Quantity:
1 Set
Catalog No. Quantity
S98668 1 Set
1 options
WARNING: This product can expose you to chemicals including Phenolphthalein and Methanol/Methyl Isobutyl Ketone, which are known to the State of California to cause cancer and reproductive harm.
Catalog No. S98668 Supplier Innovating Science™ Supplier No. IS5012
Add to Cart
Add to Cart
WARNING: This product can expose you to chemicals including Phenolphthalein and Methanol/Methyl Isobutyl Ketone, which are known to the State of California to cause cancer and reproductive harm.
Determine the pH of solutions within a wide range using this pH indicator set.
Kit Includes:
  • 30 mL Phenolphthalein, 1% Solution
  • 30 mL Universal Indicator
  • 30 mL Bromothymol Blue, 0.04%
Specifications
Quantity 1 Set
Certifications/Compliance 49 CFR 173.4
Includes 30mL phenolphthalein, 1% solution, 30 mL universal indicator, 30 mL 0.04% bromothymol blue
Volume (Metric) 3 x 30 mL
Sterility Non-sterile
Product Type pH Indicator Set