Learn More
Innovating Science™ Acid Fast Stain Chemicals Kit
GREENER_CHOICESCI_ED
Catalog No. S98662 Shop All Aldon Products
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
S98662 1 Kit
1 options
WARNING: This product can expose you to chemicals including Methanol/ Methyl Isobutyl Ketone, which is known to the State of California to cause cancer and reproductive harm.
Catalog No. S98662 Supplier Innovating Science™ Supplier No. IS5006
Add to Cart
Add to Cart
WARNING: This product can expose you to chemicals including Methanol/ Methyl Isobutyl Ketone, which is known to the State of California to cause cancer and reproductive harm.
Used to identify bacteria by staining the acid fast mycobacterium bright red and putting them against a blue background.
30 mL Methylene Blue Chloride 30 mL Carbol Fuchsin 30 mL Hydrochloric Acid, 1%
Specifications
Quantity 1 Kit
Certifications/Compliance 49 CFR 173.4
Includes 30mL methylene blue chloride, 30mL 1% alcohol solution, 30mL Carbol Fuchsin hydrochloric acid, 30mL 1% alcohol solution
Stain Type Acid Fast
Volume (Metric) 3 x 30 mL
Sterility Non-sterile
Product Type Acid Fast Stain