Learn More
Lakeland Industries Chemical Suits
Catalog No. 502840621
Encompass_Preferred
Lakeland ChemMax 4 Plus products are engineered with an expanded six-layer protective system for advanced chemical protection in the most challenging and hazardous environments. Offered in sizes LG-4X, this ChemMax 4 Plus encapsulated suit featured heat-sealed seams and can be purchase in single units. All ChemMax 4 Plus products are supported by PermaSURE - a mobile application that calculates data-backed safe-wear times. Use this tool alongside permeation breakthrough results to properly evaluate chemical suits for your employees.
Catalog No. 50-284-0621 Supplier Lakeland Industries Supplier No. C4T400YXL
Lakeland ChemMax 4 Plus products are engineered with an expanded six-layer protective system for advanced chemical protection in the most challenging and hazardous environments. Offered in sizes LG-4X, this ChemMax 4 Plus encapsulated suit featured heat-sealed seams and can be purchase in single units. All ChemMax 4 Plus products are supported by PermaSURE - a mobile application that calculates data-backed safe-wear times. Use this tool alongside permeation breakthrough results to properly evaluate chemical suits for your employees.