Learn More
Lakeland Industries Chemical Suits
Catalog No. 502840658
Encompass_Preferred
Lakeland's ChemMax 2 chemical suit with heat-sealed seams features a Saranex 23P film on two layers of bicomponent spun bound non-woven substrate. Ideal for chemical mixing and handling, environmental clean-up, and hazardous materials remediation. This configuration features an attached hood and boots. Offered in cases of 6 each, C2T151 chemical coverall is offered in sizes MD-4X.
Catalog No. 50-284-0658 Supplier Lakeland Industries Supplier No. C2T1514X
Lakeland's ChemMax 2 chemical suit with heat-sealed seams features a Saranex 23P film on two layers of bicomponent spun bound non-woven substrate. Ideal for chemical mixing and handling, environmental clean-up, and hazardous materials remediation. This configuration features an attached hood and boots. Offered in cases of 6 each, C2T151 chemical coverall is offered in sizes MD-4X.