Learn More
Lakeland Industries Chemical Suits
Catalog No. 502840702
Encompass_Preferred
Multicapa tissue with a high level of protection. Fabor weight: 365 GSM. Design perfected with doubly sealed seams (inside and outside). Standard or wide field facial protector options; Two -layer facial protector with exclusive sealing technology to offer a high chemical barrier. Double layer glove system with protection against chemical substances: Interior chemical glove with 27 mm butyl outlet glove. Tested in front of a wide range of chemical substances for military purposes in anti -terrorist and civil defense operations. Extremely soft and flexible fabric for comfort. Design options with front or rear input. Two exhaust valves integrated in the rear. Boot with integrated sock and protective flaps on boots. All interceptor costumes are subjected to gas tank tests before leaving the factory. Gas tightness test kits available separately.
Catalog No. 50-284-0702 Supplier Lakeland Industries Supplier No. INT640WB2X
Add to Cart
Add to Cart
Multicapa tissue with a high level of protection. Fabor weight: 365 GSM. Design perfected with doubly sealed seams (inside and outside). Standard or wide field facial protector options; Two -layer facial protector with exclusive sealing technology to offer a high chemical barrier. Double layer glove system with protection against chemical substances: Interior chemical glove with 27 mm butyl outlet glove. Tested in front of a wide range of chemical substances for military purposes in anti -terrorist and civil defense operations. Extremely soft and flexible fabric for comfort. Design options with front or rear input. Two exhaust valves integrated in the rear. Boot with integrated sock and protective flaps on boots. All interceptor costumes are subjected to gas tank tests before leaving the factory. Gas tightness test kits available separately.