Learn More
Lakeland Industries Chemical Suits
Supplier:  Lakeland Industries INT650BXL
Encompass_Preferred
Interceptor - Multi-layer, High Barrier Fully Encapsulating Gas Tight Type 1a Suit has been tested against a full range of chemical warfare agents for anti-terror and civil defence operations. Very soft and flexible material for enhanced comfort.\n \n Multi-layer, high barrier fabric \n Fabric weight 365gsm \n Superior design featuring double-taped seams (inside & out) \n Standard or wide-vision visor options; two-layer visor with unique sealing technology for high chemical barrier \n Double layer chemical glove system: inner chemical glove with outer barrier 27mil butyl glove \n Tested against a full range of chemical warfare agents for anti-terror and civil defence operations. Very soft and flexible material for enhanced comfort. \n Two rear mounted exhaust valves \n Interceptor Training suits (INT497 Front Entry / INT491 Rear Entry) and Gas-tight test kits are available separately \n
Catalog No. 50-284-0722