Learn More
Lakeland Industries Chemical Suits
Catalog No. 502840506
Encompass_Preferred
Lakeland's ChemMax 2 chemical apron with bound seams is ideal for chemical mixing and handling, environmental clean-up, and hazardous materials remediation. The full sleeve and bound seam features offer additional protection against liquid splash and spray. All Lakeland ChemMax 2 products use a Saranex 23P coating for moderate chemical protection. Sold in cases of 50, this chemical-resistant apron is offered in sizes SM-4X.
Catalog No. 50-284-0506 Supplier Lakeland Industries Supplier No. C2B657LG
Lakeland's ChemMax 2 chemical apron with bound seams is ideal for chemical mixing and handling, environmental clean-up, and hazardous materials remediation. The full sleeve and bound seam features offer additional protection against liquid splash and spray. All Lakeland ChemMax 2 products use a Saranex 23P coating for moderate chemical protection. Sold in cases of 50, this chemical-resistant apron is offered in sizes SM-4X.