Learn More
Lakeland Industries Chemical Suits
Catalog No. 502840763
Encompass_Preferred
Lakeland's ChemMax 1 chemical boot covers are designed with serged seams and an elastic top for additional security at the interface points. The 17" covers are offered in cases with 200 pairs/case with sizes ranging from MD-2XL. Ideal for general industry, environmental clean-up, and chemical handling.
Catalog No. 50-284-0763 Supplier Lakeland Industries Supplier No. C1S903YPXL
Lakeland's ChemMax 1 chemical boot covers are designed with serged seams and an elastic top for additional security at the interface points. The 17" covers are offered in cases with 200 pairs/case with sizes ranging from MD-2XL. Ideal for general industry, environmental clean-up, and chemical handling.