Learn More
Lakeland Industries Chemical Suits
Catalog No. 502840765
Encompass_Preferred
Lakeland's ChemMax 1 chemical suit with heat-sealed seams provides you with defense against light liquid splash in general industry, environmental cleaning, and chemical handling applications. This chemical coverall features an attached hood and boots. All ChemMax 1 products are constructed with a unique polyethylene barrier film that provides you with protection from harmful contaminants. Offered in cases of 6 each, C1T150Y is offered in sizes MD-4X.
Catalog No. 50-284-0765 Supplier Lakeland Industries Supplier No. C1T150YLG
Lakeland's ChemMax 1 chemical suit with heat-sealed seams provides you with defense against light liquid splash in general industry, environmental cleaning, and chemical handling applications. This chemical coverall features an attached hood and boots. All ChemMax 1 products are constructed with a unique polyethylene barrier film that provides you with protection from harmful contaminants. Offered in cases of 6 each, C1T150Y is offered in sizes MD-4X.