Learn More
Lakeland Industries Chemical Suits
Catalog No. 502840993
Encompass_Preferred
Lakeland Interceptor Plus, Level A suits are gas tight and are the ultimate line of chemical protection. INT640B front entry suits are configured with gloves, boots, face shield, and heat-sealed seams. Available in sizes SM-5X, these protective suits are ideal when the maximum level of chemical protection is needed, like emergency response and HazMat operations.
Catalog No. 50-284-0993 Supplier Lakeland Industries Supplier No. INT640BLG
Lakeland Interceptor Plus, Level A suits are gas tight and are the ultimate line of chemical protection. INT640B front entry suits are configured with gloves, boots, face shield, and heat-sealed seams. Available in sizes SM-5X, these protective suits are ideal when the maximum level of chemical protection is needed, like emergency response and HazMat operations.