Learn More
Lakeland Industries Chemical Suits
Catalog No. 502840995
Encompass_Preferred
Multi-layer, high barrier fabric Fabric weight 365gsm Superior design featuring double-taped seams (inside & out) WIDE two-layer visor design with unique sealing technology for high chemical barrier Double layer chemical glove system: inner chemical glove with outer barrier 27mil butyl glove Tested against a full range of chemical warfare agents for anti-terror and civil defence operations Very soft and flexible material for enhanced comfort REAR entry desig To rear mounted exhaust valve Interceptor Training Suits (INT497 Front Entry / INT491 Rear Entry) and gas tight test kits are available separately
Catalog No. 50-284-0995 Supplier Lakeland Industries Supplier No. INT650WB3X
Multi-layer, high barrier fabric Fabric weight 365gsm Superior design featuring double-taped seams (inside & out) WIDE two-layer visor design with unique sealing technology for high chemical barrier Double layer chemical glove system: inner chemical glove with outer barrier 27mil butyl glove Tested against a full range of chemical warfare agents for anti-terror and civil defence operations Very soft and flexible material for enhanced comfort REAR entry desig To rear mounted exhaust valve Interceptor Training Suits (INT497 Front Entry / INT491 Rear Entry) and gas tight test kits are available separately