Learn More
Lakeland Industries Chemical Suits
Catalog No. 502840519
Encompass_Preferred
Lakeland's ChemMax 1 chemical coverall with bound seams provides you with defense against light liquid splash in general industry, environmental cleaning, and chemical handling applications. This configuration features an attached hood. All ChemMax 1 products are constructed with a unique polyethylene barrier film that provides you with protection from harmful contaminants. Offered in cases of 25 each, you can purchase C1B428Y in sizes MD-4X.
Catalog No. 50-284-0519 Supplier Lakeland Industries Supplier No. C1B428YLG
Lakeland's ChemMax 1 chemical coverall with bound seams provides you with defense against light liquid splash in general industry, environmental cleaning, and chemical handling applications. This configuration features an attached hood. All ChemMax 1 products are constructed with a unique polyethylene barrier film that provides you with protection from harmful contaminants. Offered in cases of 25 each, you can purchase C1B428Y in sizes MD-4X.