Learn More
Lakeland Industries Chemical Suits
Catalog No. 502840546
Encompass_Preferred
Lakeland's ChemMax 1 chemical coverall with bound seams provides you with defense against light liquid splash in general industry, environmental cleaning, and chemical handling applications. This configuration features an attached hood and boots. All ChemMax 1 products are constructed with a unique polyethylene barrier film that provides you with protection from harmful contaminants. Offered in cases of 25 each, you can purchase C1B414Y in sizes MD-4X.
Catalog No. 50-284-0546 Supplier Lakeland Industries Supplier No. C1B414Y2X
Lakeland's ChemMax 1 chemical coverall with bound seams provides you with defense against light liquid splash in general industry, environmental cleaning, and chemical handling applications. This configuration features an attached hood and boots. All ChemMax 1 products are constructed with a unique polyethylene barrier film that provides you with protection from harmful contaminants. Offered in cases of 25 each, you can purchase C1B414Y in sizes MD-4X.