Learn More
Lakeland Industries Chemical Suits
Catalog No. 502840552
Encompass_Preferred
Lakeland's ChemMax 1 chemical coverall with serged seams provides you with defense against light liquid splash in general industry, environmental cleaning, and chemical handling applications. This configuration features an attached hood and boots. All ChemMax 1 products are constructed with a unique polyethylene barrier film that provides you with protection from harmful contaminants. Offered in cases of 25 each, you can purchase C1S414Y in sizes SM-5XL.
Catalog No. 50-284-0552 Supplier Lakeland Industries Supplier No. C1S414Y6X
Lakeland's ChemMax 1 chemical coverall with serged seams provides you with defense against light liquid splash in general industry, environmental cleaning, and chemical handling applications. This configuration features an attached hood and boots. All ChemMax 1 products are constructed with a unique polyethylene barrier film that provides you with protection from harmful contaminants. Offered in cases of 25 each, you can purchase C1S414Y in sizes SM-5XL.