Learn More
Lakeland Industries Chemical Suits
Catalog No. 502840823
Encompass_Preferred
Lakeland's ChemMax 2 chemical coverall with bound seams features a Saranex 23P film on two layers of bicomponent spun bound nonwoven substrate. Ideal for chemical mixing and handling, environmental clean-up, and hazardous materials remediation. This configuration features an attached hood. Sold in cases of 12, this chemical suit is offered in sizes MD-4X.
Catalog No. 50-284-0823 Supplier Lakeland Industries Supplier No. C2B4286X
Lakeland's ChemMax 2 chemical coverall with bound seams features a Saranex 23P film on two layers of bicomponent spun bound nonwoven substrate. Ideal for chemical mixing and handling, environmental clean-up, and hazardous materials remediation. This configuration features an attached hood. Sold in cases of 12, this chemical suit is offered in sizes MD-4X.