Learn More
Lakeland Industries Chemical Suits
Catalog No. 502840641
Encompass_Preferred
Durable and lightweight, Lakeland ChemMax 3 uses innovative materials and design for a superior chemical coverall. This heat-sealed seam coverall with attached hood provides a barrier against a broad spectrum of toxic industrial chemicals, dual-use chemicals, chemical warfare agents, and other harmful contaminants. For all ChemMax 3 products, the unique barrier film is significantly softer than other chemical suits on the market, resulting in a quieter, more comfortable garment. All ChemMax 3 products are supported by PermaSURE - a mobile application that calculates data-backed safe-wear times. Use this tool alongside permeation breakthrough results to properly evaluate chemical suits for your employees. This chemical coverall is sold in cases with 6 garments and is offered in sizes MD-4X.
Catalog No. 50-284-0641 Supplier Lakeland Industries Supplier No. C3T132XL
Durable and lightweight, Lakeland ChemMax 3 uses innovative materials and design for a superior chemical coverall. This heat-sealed seam coverall with attached hood provides a barrier against a broad spectrum of toxic industrial chemicals, dual-use chemicals, chemical warfare agents, and other harmful contaminants. For all ChemMax 3 products, the unique barrier film is significantly softer than other chemical suits on the market, resulting in a quieter, more comfortable garment. All ChemMax 3 products are supported by PermaSURE - a mobile application that calculates data-backed safe-wear times. Use this tool alongside permeation breakthrough results to properly evaluate chemical suits for your employees. This chemical coverall is sold in cases with 6 garments and is offered in sizes MD-4X.