Learn More
Thermo Scientific CMX25, Cell Culture Flask, 25 cm2, Vented Cap, Sterile
Catalog No. NC1320100 Shop All Thermo Scientific Products
Encompass_Preferred
The Thermo Scientific CMX25 is a compact, portable CO2 incubator designed for use in IVF clinics and laboratories. It features a 25 L capacity stainless steel chamber, IR CO2 sensor, adjustable temperature from ambient +5°C to 50°C, humidity control from Ambient to 95% RH. The unit provides a stable, consistent environment for culturing cells and tissues outside of a traditional CO2 incubator. Its small footprint and quiet operation allow it to be conveniently used directly on the benchtop in close proximity to other lab equipment. The CMX25 requires minimal maintenance and its seamless, stainless steel chamber is easy to clean.
Catalog No. NC1320100 Supplier Thermo Scientific Supplier No. CMX25
The Thermo Scientific CMX25 is a compact, portable CO2 incubator designed for use in IVF clinics and laboratories. It features a 25 L capacity stainless steel chamber, IR CO2 sensor, adjustable temperature from ambient +5°C to 50°C, humidity control from Ambient to 95% RH. The unit provides a stable, consistent environment for culturing cells and tissues outside of a traditional CO2 incubator. Its small footprint and quiet operation allow it to be conveniently used directly on the benchtop in close proximity to other lab equipment. The CMX25 requires minimal maintenance and its seamless, stainless steel chamber is easy to clean.