Learn More
Neta Scientific Agilent GC Split Vent Replacement Cartridge,2/pk
SDP
Catalog No. 50924558
Encompass_Preferred
This item is manufactured by Agilent
Agilent GC Split Vent Replacement Cartridge,2/pk?Replacement Filter Cartridge Kit for?G1544-60610 (2/pkg);?Kit contains 2 replacement split vent?cartridges with 4 o-rings
- AA
Catalog No. 50-924558 Supplier Neta Scientific Supplier No. G154480530
This item is manufactured by Agilent
Agilent GC Split Vent Replacement Cartridge,2/pk?Replacement Filter Cartridge Kit for?G1544-60610 (2/pkg);?Kit contains 2 replacement split vent?cartridges with 4 o-rings
- AA