Learn More
FUJIFILM Biosciences Chemically Dfined animal 1L
Catalog No. 502892204
Encompass_Preferred
Designed for use in NK and NKT cell-based immunotherapy research and translational applications, this advanced formulation delivers growth while maintaining NK and NKT cell functionality and potency - without the need for serum supplementation.Provides Optimal PerformanceChemically defined, animal component-free natural killer cell culture mediumSupports robust cell expansion and viability in heterogeneous PBMC culture enriched for CD56+ cellsSupports expansion of NK cells when supplemented with IL-2, IL-12, IL-18, and IL-21Supports NKT cell expansion when supplemented with IL-2Maintains high NK and NKT cytotoxic capabilityDesigned and manufactured to facilitate transfer from research to clinic. Supported by robust raw material controls and supply chain management
Catalog No. 50-289-2204 Supplier FUJIFILM Biosciences Supplier No. 912151L
Designed for use in NK and NKT cell-based immunotherapy research and translational applications, this advanced formulation delivers growth while maintaining NK and NKT cell functionality and potency - without the need for serum supplementation.Provides Optimal PerformanceChemically defined, animal component-free natural killer cell culture mediumSupports robust cell expansion and viability in heterogeneous PBMC culture enriched for CD56+ cellsSupports expansion of NK cells when supplemented with IL-2, IL-12, IL-18, and IL-21Supports NKT cell expansion when supplemented with IL-2Maintains high NK and NKT cytotoxic capabilityDesigned and manufactured to facilitate transfer from research to clinic. Supported by robust raw material controls and supply chain management