Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Chemicon™ Dextran Sulfate 50% Solution

Catalog No. S4030 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 mL
200 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S4030 100 mL
S4031 200 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S4030 Supplier MilliporeSigma™ Supplier No. S4030
Add to Cart
Add to Cart

Used as additive in probe hybridization solutions.

  • Presence accelerates rate of hybridization by 10 fold
  • Favors formation of probe networks
  • Greater sensitivity during membrane bound nucleic acid analysis
  • 50% (w/v) solution
TRUSTED_SUSTAINABILITY

Specifications

Includes Mitochondria Extraction Buffer, Cytosol Extraction Buffer, Protease Inhibitor Cocktail, Reducing Reagent, and user protocol.
Quantity 100 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.