Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Chemicon™ Embryonic Stem Cell Marker Sample Kit

Catalog No. SCR002 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
Sufficient for 20 Reactions (including controls)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SCR002 Sufficient for 20 Reactions (including controls)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SCR002 Supplier MilliporeSigma™ Supplier No. SCR002

Contains monoclonal antibodies for the detection of the cell-surface Stage-Specific Embryonic Antigens (SSEA-1, SSEA-3 & SSEA-4), expression of TRA-1-60, TRA-1-81 antigens and Oct-4. Chemicon™ Embryonic Stem Cell Marker Sample Kit includes six monoclonal antibodies for the analysis of ES cell marker expression.

  • Species Reactivity: Human, Mouse
TRUSTED_SUSTAINABILITY

Specifications

Cell Type Human Embryonic Stem Cells, Mouse Embryonic Stem Cells, Induced Pluripotent Stem Cells
Product Type Stem Cell Marker Sample Kit
For Use With (Application) Immunocytochemistry
Quantity Sufficient for 20 Reactions (including controls)
Regulatory Status RUO
Content And Storage
  • MS X SSEA-1, IgM, clone MC-480: (Part No. MAB4301-20). One vial containing 20μL of 1mg/mL monoclonal antibody.
  • RT X SSEA-3, IgM, clone MC-631: (Part No. MAB4303-20). One vial containing 20μL of 1mg/mL monoclonal antibody.
  • MS X SSEA-4, IgG, clone MC-813-70: (Part No. MAB4304-20). One vial containing 20μL of 1mg/mL monoclonal antibody.
  • MS X TRA-1-60, IgM, clone TRA-1-60: (Part No. MAB4360-20). One vial containing 20μL of 1mg/mL monoclonal antibody
  • MS X TRA-1-81, IgM, clone TRA-1-81: (Part No: MAB4381-20). One vial containing 20μL of 1mg/mL monoclonal antibody.
  • MS X Oct-4, IgG, clone 9E3.2: (Part number: MAB4305-20). One vial containing 20μL of 1mg/mL monoclonal antibody.
When stored at 2 to 8º C, the kit components are stable up to the expiration date. Do not freeze or expose to elevated temperatures. Discard any remaining reagents after the expiration date.

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.