Learn More
MilliporeSigma™ Human STEMCCA Constitutive (OKSM) Lentivirus
Catalog No. SCR5441 Shop All MilliporeSigma Products
Change view
Click to view available options
Human STEMCCA Constitutive (OKSM) Lentivirus
Catalog No.
SCR5441 Human STEMCCA Constitutive (OKSM) Lentivirus
1 options
Catalog No. SCR5441 Supplier MilliporeSigma™ Supplier No. SCR5441
For use with Human STEMCCA Constitutive Polycistronic (OKSM) Lentivirus Reprogramming Kit (SCR544).
  • Available with Human STEMCCA Constitutive Polycistronic (OKSM) Lentivirus Reprogramming Kit (SCR544) as part of Kit component
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Lentivirus is stable for at least 6 months when stored at -80°C. After first thaw, place immediately on ice and store in working aliquots to avoid further freeze thaws. Avoid freeze thaws as this will result in a decrease in the virus titer. Polybrene transfection reagent is stable for at least 1 year when stored at -20°C.
Product Type Lentivirus Reprogramming Kit