Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Chemicon™ ENStem-A™ Human Neural Progenitor Cells

Catalog No. SCC003 Shop All MilliporeSigma Products
Change view
Click to view available options
Cell Type:
Human Neural Progenitor Cells
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Cell Type
SCC003 Human Neural Progenitor Cells
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SCC003 Supplier MilliporeSigma™ Supplier No. SCC003
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For use with ENStem-A Neural Freezing Medium (SCM011). MilliporeSigma™; Chemicon™ ENStem-A™ Human Neural Progenitor Cells is cultured in defined serum-free conditions.

  • Applications: Stem cell culture
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at -20°C. Refer to lot expiration date on label. Used with ENStem-A Neural Freezing Medium (SCM011)
Cell Type Human Neural Progenitor Cells
For Use With (Application) Stem cell culture
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.