Learn More
MilliporeSigma™ Chemicon™ Human iPS Selection Kit
Catalog No. SCR502 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Kit (Sufficient for 40 Tests/Antibody)
Catalog No. Quantity
SCR502 1 Kit (Sufficient for 40 Tests/Antibody)
1 options
Catalog No. SCR502 Supplier MilliporeSigma™ Supplier No. SCR502
Add to Cart
Add to Cart
Allows live cell imaging and identification of fully reprogrammed human induced pluripotent Stem cells from heterogeneous population of reprogramming intermediates. MilliporeSigma™; Chemicon™ Human iPS Selection Kit is a quick, easy and non-invasive method to monitor the pluripotent state of live human ESC and fully reprogrammed human iPS cells using immunocytochemistry (ICC).
  • Enables live selection of human iPS cells that can be further passaged and expanded for downstream applications
  • Selected live stained human iPS clones maintain normal human ESC morphology and can proliferate and be passaged normally
  • Fully reprogrammed human iPS cells possess following characteristics:
  • They downregulate expression of fibroblast marker, CD13
  • They upregulate expression of pluripotent markers, SSEA-4 and TRA-1-60
  • They silence viral transgenes
  • Activates endogenous expression of Nanog
  • Assume HoeschstDim phenotype
  • Kit Components:
  • Anti-SSEA-4 PE (Mouse monoclonal IgG3), Cat. No. CS204438. One vial containing 50μg of 0.25mg/mL purified mouse IgG antibody
  • Anti-TRA-1-60 FITC (Mouse monoclonal IgM), Cat. No. CS204460. One vial containing 50μg of 0.5mg/mL purified mouse IgM antibody
  • Anti-CD13 PE (Mouse monoclonal IgG1), Cat. No. CS204449. One vial containing 50μg of 1mg/mL purified mouse IgG antibody
  • Hoechst Nuclear Stain, Cat. No. CS204269. One vial containing 1μL of 1mg/mL stock solution
TRUSTED_SUSTAINABILITY
Specifications
Product Type Human iPS Selection Kit
For Use With (Application) Immunocytochemistry
Quantity 1 Kit (Sufficient for 40 Tests/Antibody)
Regulatory Status RUO
Content And Storage Stable for 6 months at 2°to 8°C from date of receipt. Handling Recommendations: Upon first thaw, and prior to removing the cap, centrifuge the vial and gently mix the solution. Maintain refrigerated at 2° to 8°C, protected from light in undiluted aliquots.