Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Chemicon™ Human Mesenchymal Stem Cell Characterization Kit

Catalog No. SCR067 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SCR067 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SCR067 Supplier MilliporeSigma™ Supplier No. SCR067
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Contains panel of positive and negative selection markers for characterization of mesenchymal stem cell population in human samples. MilliporeSigma™; Chemicon™ Human Mesenchymal Stem Cell Characterization Kit is used in Immunocytochemistry and stem cell culture applications.

  • Positive cell markers include antibodies directed against cell-surface molecules present on mesenchymal stem cells: CD44, CD90, STRO-1, and CD146
  • Two specific hematopoeitic cell surface markers are provided whose expressions should not be present on mesenchymal stem cells: CD14 (present on leukocytes) and CD19 (present on B-lymphocytes)
  • Components:
  • Mouse anti-HCAM
  • Mouse anti-THY-1 (CD90)
  • Mouse anti-STRO-1
  • Mouse anti-MCAM (CD146)
  • Mouse anti-CD19
  • Mouse anti-CD14
TRUSTED_SUSTAINABILITY

Specifications

Product Type Stem Cell Characterization Kit
For Use With (Application) Immunocytochemistry, Stem Cell Culture
Quantity 1 Kit
Regulatory Status RUO
Content And Storage When stored at the recommended storage conditions (refer to Kit Components), components are stable up to the expiration date. Do not expose to elevated temperatures. Discard any remaining reagents after the expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.