Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Chemicon™ Human Oligodendrocyte Progenitors (OPCs)

Catalog No. CS204496 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500,000 viable cells
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CS204496 500,000 viable cells
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CS204496 Supplier MilliporeSigma™ Supplier No. CS204496
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For use with Human Oligodendrocyte Differentiation Kit (SCR600). MilliporeSigma™; Chemicon™ Human Oligodendrocyte Progenitors (OPCs) is used in expansion and spontaneous differentiation into mature oligodendrocytes.

  • Derived from NIH approved H9 human embryonic stem cells (hESCs)
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Component of Human Oligodendrocyte Differentiation Kit (SCR600)
Cell Type Human Oligodendrocyte Progenitors (OPCs)
For Use With (Application) Stem cell culture
Quantity 500,000 viable cells
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.