Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Chemicon™ Laminin, Mouse Purified

Catalog No. p-5394010 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC095 1 mg
CC0955MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC095 Supplier MilliporeSigma™ Supplier No. CC095
Only null left
Add to Cart
Add to Cart

For Cell Culture applications

TRUSTED_SUSTAINABILITY

Specifications

Concentration 1 mg/mL
For Use With (Application) Cell Culture
Formulation Liquid; in 0.05 M Tris-HCl, pH 7.4, containing 0.15 M NaCl. 
Quantity 1 mg
Source Isolated from Engelbreth-Holm-Swarm (EHS) mouse sarcoma
Product Type Laminin, Mouse Purified
Content And Storage Store at -20°C for up to six months. 
Form Liquid
Purity or Quality Grade >95% by SDS-PAGE 
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.