Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Chemicon™ Mesenchymal Adipogenesis Kit

Catalog No. SCR020 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SCR020 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SCR020 Supplier MilliporeSigma™ Supplier No. SCR020
Add to Cart
Add to Cart

Obtains 30% mature adipocytes from the rat bone marrow derived mesenchymal stem cells.

  • Stem Cell Type: Mesenchymal Stem Cells
TRUSTED_SUSTAINABILITY

Specifications

Cell Type Mesenchymal Stem Cells
Product Type Stem Cell Adipogenesis Kit
Quantity 1 Kit
Regulatory Status RUO
Content And Storage
  • Dexamethasone Solution: (Catalog No. 90357) One vial containing 100μL of 10mM Dexamethasone in ethanol. Store at -20°C.
  • IBMX Solution: (Catalog No. 90355) One vial containing 250μL of 0.5M 3-isobutyl-1-methylxanthine (IBMX) in DMSO. Store at -20°C.
  • Insulin Solution: (Catalog No. 90356) Two vials containing 250μL of 10mg/mL recombinant human insulin. Store at -20°C.
  • Indomethacin Solution: (Catalog No. 2003626) Three vials containing 1mL of 10mM Indomethacin in methanol. Store at -20°C.
  • Oil Red O Solution: (Catalog No. 90358) One bottle containing 60mL 0.36% Oil Red O Solution in 60% isopropanol. Store at room temperature.
  • Hematoxylin Solution: (Catalog No. 2003732) One bottle containing 50mL Hematoxylin. Store at room temperature.
Note: Kit components require two different storage temperatures. 
Dexamethasone Solution, IBMX Solution, Insulin Solution and Indomethacin Solution should be stored at -20°C. Oil Red O and Hematoxylin Solutions should be stored at room temperature. Storage of Oil Red O Solution at -20°C may result in formation of insoluble precipitates and is not recommended. If Oil Red O solution forms a precipitate, remove particulates by passage through a 0.22 or 0.45-micron filter. 
Precautions: Oil Red O stains skin and clothing. IBMX, Dexamethasone and Indomethacin are irritants and potentially toxic. DMSO is readily absorbed through the skin. Wear a lab coat and gloves when handling these solutions. Isopropanol is flammable. Keep solutions containing isopropanol (Oil Red O Solution) away from open flames.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.