Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Chemicon™ Mesenchymal Stem Cell Osteogenesis Kit

Catalog No. SCR028 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SCR028 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SCR028 Supplier MilliporeSigma™ Supplier No. SCR028
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Promotes osteogenic differentiation of mesenchymal stem cells. Contains all the reagents necessary to readily differentiate mesenchymal stem cells to an osteogenic lineage as assessed by Alizarin Red staining.

  • Detection method: Colorimetric
TRUSTED_SUSTAINABILITY

Specifications

Product Type Stem Cell Osteogenesis Kit
Quantity 1 Kit
Regulatory Status RUO
Content And Storage
  • Dexamethasone Solution: (Catalog No. 90357) One vial containing 100μL of 10mM dexamethasone in ethanol. Store at -20°C.
  • Ascorbic Acid 2-Phosphate Solution: (Catalog No. 2004011) One vial containing 500μL of 100μM ascorbic acid 2-phosphate in water. Store at -20°C.
  • Glycerol 2- Phosphate Solution: (Catalog No. 2004010) Three vials containing 1mL of 1M glycerol 2-phosphate in water. Store at -20°C.
  • Collagen, Type I: (Catalog No. 2004013) One vial containing 150μg collagen Type I. Store at -20°C.
  • Vitronectin: (Catalog No. 2004012) One vial containing 150μg vitronectin. Store at -20°C.
  • Alizarin Red Solution: (Catalog No. 2003999) One bottle containing 50mL Alizarin Red Solution. Store at 2° to 8°C.
Note: Kit components require two different storage temperatures.
Dexamethasone Solution, Ascorbic Acid 2-Phosphate Solution, Glycerol 2- Phosphate Solution, Collagen Type I, and Vitronectin should be stored at -20ºC. Alizarin Red Solution should be stored at 2° to 8°C. 
Precautions: Dexamethasone is an irritant and potentially toxic. DMSO is readily absorbed through the skin. Wear a lab coat and gloves when handling these solutions. Please refer to the Material Safety Data Sheet at for further precautions.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.