Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Chemicon™ Millicoat™ Human Laminin Coated Strips (96-Wells)

Catalog No. ECM103 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 x 96-well Plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ECM103 1 x 96-well Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ECM103 Supplier MilliporeSigma™ Supplier No. ECM103
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For enzyme assay applications

  • Provided as 12 x 8-well removable strips in a plate frame for convenience and flexibility in designing assays
  • Wells in rows A to G have been coated with Human Laminin
  • Row H of each strip is coated with BSA which serves as negative assay control
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Enzyme Assay
Quantity 1 x 96-well Plate
Product Type Human Laminin Coated Strips
Content And Storage May be stored at 2° to 8°C in the foil pouch for at least 3 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.