Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Chemicon™ Quantitative Alkaline Phosphatase ES Characterization Kit

Catalog No. SCR066 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 Assays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SCR066 100 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SCR066 Supplier MilliporeSigma™ Supplier No. SCR066
Add to Cart
Add to Cart

Quantifies amount of alkaline phosphatase present within embryonic stem cell cultures using convenient 96-well colorimetric enzymatic assay. MilliporeSigma™; Chemicon™ Quantitative Alkaline Phosphatase ES Characterization Kit is used in Stem Cell Culture applications.

  • Components: 
  • p-NPP Substrate Concentrate (50X): (Part No. ES008-200UL) 200μL
  • p-NPP Buffer: (Part No. ES011-10ML) 10mL
  • Reaction Stop Solution: (Part No.CS200653) 5mL
  • 1X Wash Solution: (Part No. CS200652) 2 X 125mL
  • Recombinant Alkaline Phosphatase Standard: (Part No. CS200654) 0.5μg (10μg/mL)
TRUSTED_SUSTAINABILITY

Specifications

Product Type Stem Cell Characterization Kit
For Use With (Application) Stem Cell Culture
Quantity 100 Assays
Regulatory Status RUO
Content And Storage All reagents are to be stored at 2 to 8°C until expiration date. p-NPP substrate concentrate and p-NPP buffer are light sensitive and should not be exposed to light. Reaction stop solution contains 1N sodium hydroxide and is corrosive. Avoid skin contact and inhalation.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.