Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Chemicon™ Rat Mesenchymal Stem Cells (Bone Marrow)

Catalog No. SCR027 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10e6 cells
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SCR027 10e6 cells
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SCR027 Supplier MilliporeSigma™ Supplier No. SCR027
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Isolated from iliac crest of adult Fisher 344 rats provide convenient source of multipotent rat mesenchymal stem cells

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in liquid nitrogen
Cell Type Rat Mesenchymal Stem Cells (RMSC, Bone Marrow)
For Use With (Application) Stem cell culture
Quantity 10e6 cells

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.