Learn More
Bio Rad Laboratories CHEMIDOC IMAGING SYSTEM
Catalog No. NC2005948
Encompass
  • Imaging modes include chemiluminescence, colorimetric, gel documentation, and stain-free.
  • Supports applications for DNA/protein gel imaging and chemiluminescent western blot detection.
  • Compatible with stains such as ethidium bromide, SYPRO Ruby, Coomassie, and silver stains.
  • Utilizes stain-free technology for protein normalization.
  • Features a 12 in touch-screen display for user-friendly operation.
  • Includes Image Lab Touch Software and Image Lab Software for data analysis.
  • Smart Tray Technology™ enables automatic recognition of application-specific trays.
  • Precalibrated autofocus enhances precision in imaging.
  • Auto-exposure options available in rapid or optimal modes.
  • Employs image flat fielding for image correction.
Catalog No. NC2005948 Supplier Bio Rad Laboratories Supplier No. 12003153
Add to Cart
Add to Cart
Request a Quote
  • Imaging modes include chemiluminescence, colorimetric, gel documentation, and stain-free.
  • Supports applications for DNA/protein gel imaging and chemiluminescent western blot detection.
  • Compatible with stains such as ethidium bromide, SYPRO Ruby, Coomassie, and silver stains.
  • Utilizes stain-free technology for protein normalization.
  • Features a 12 in touch-screen display for user-friendly operation.
  • Includes Image Lab Touch Software and Image Lab Software for data analysis.
  • Smart Tray Technology™ enables automatic recognition of application-specific trays.
  • Precalibrated autofocus enhances precision in imaging.
  • Auto-exposure options available in rapid or optimal modes.
  • Employs image flat fielding for image correction.