Learn More
Perkin Elmer US LLC Chemifold Mixing Manifold for Glass Gas-Liquid Separator
Catalog No. 509042308
Encompass_Preferred
This Chemifold Mixing Manifold is compatible with glass gas-liquid separators of flow injection systems such as the FIAS and FIMS instruments.
Catalog No. 50-904-2308 Supplier Perkin Elmer US LLC Supplier No. B0187258
This Chemifold Mixing Manifold is compatible with glass gas-liquid separators of flow injection systems such as the FIAS and FIMS instruments.