Learn More
Hoefer, Inc Chemi FX9 imaging system ( Chemi+Gel+RGB)
SDP
Supplier:  Hoefer, Inc CHEMI FX9 IMAGING SYSTEM
Encompass_Preferred
"The perfect gel documentation system for detailed image analysisLooking for a Western blot imaging system that gives you more control over how you detect proteins? Look no further than the Chemi FX6/FX9. This powerful system, powered by Hoefer’s GeneSys software, lets you choose from a variety of chemiluminescence and fluorescence reagents from any manufacturer so that you can detect your proteins your way. With a 6 or 9 megapixel camera and HI LED lighting options, you’ll get sensitive, publication-quality images with minimal background interference. Plus, the system has unlimited copies of GeneTools analysis software to help you maximise your data.The Chemi FX6/FX9 is the perfect system for gel documentation, imaging and analysis in your lab. The in-house designed, FREE software with unlimited licences ensures you can detect chemiluminescence effectively and efficiently.
Catalog No. NC3655222
Add to Cart
Request a Quote