Learn More
Crescent Chemical Co Inc CHEMILUMINESCENCE REAGENT
SDP
Catalog No. 50248625
Encompass_Preferred
Chemiluminescence reagent for Horseradish Peroxidase Ready-to-use substrate solution for chemiluminescence detection of membrane bound antigens or nucleic acid sequences directly with Horseradish Peroxidase (HRP) or indirectly with HRP-conjugated antibodies or Streptavidin
Catalog No. 50-248-625 Supplier Crescent Chemical Co Inc Supplier No. SE42582.02
Chemiluminescence reagent for Horseradish Peroxidase Ready-to-use substrate solution for chemiluminescence detection of membrane bound antigens or nucleic acid sequences directly with Horseradish Peroxidase (HRP) or indirectly with HRP-conjugated antibodies or Streptavidin