Learn More
LICORbio WesternSure Pre-stained Chemiluminescent Protein Ladder (250 L)
Catalog No. NC0986471
Encompass_Preferred
The WesternSure chemiluminescent protein ladder is the only pre-stained, multi-colored protein ladder for both film and digital chemiluminescence detection. It fits with your current protocol without optimization, does not rely on strep-tagged HRP or secondary antibody binding, works with a variety of substrates, and provides a vibrant color scheme that allows you to quickly orient your membrane
Catalog No. NC0986471 Supplier LICORbio Supplier No. 92698000
The WesternSure chemiluminescent protein ladder is the only pre-stained, multi-colored protein ladder for both film and digital chemiluminescence detection. It fits with your current protocol without optimization, does not rely on strep-tagged HRP or secondary antibody binding, works with a variety of substrates, and provides a vibrant color scheme that allows you to quickly orient your membrane