Description
- 1/16 in. fittings
- 0.010 in. ports
- Available in 6-port or 10-port configurations
- Available with manual or microelectric actuator
Specifications
Specifications
| Volume (Metric) Sample | 100 μL |