Learn More
Thermo Scientific™ Cheminert™ Sample injector loops, stainless steel, 10μL
Catalog No. 03052410 Shop All Thermo Scientific Products
Change view
Click to view available options
Volume (Metric) Sample:
10 μL
Catalog No. Volume (Metric) Sample
03-052-410 10 μL
1 options
Catalog No. 03-052-410 Supplier Thermo Scientific™ Supplier No. CSL10
Use Thermo Scientific™ Cheminert™ Model C2 Microbore Injectors as injectors or switching valves.
Use Thermo Scientific™ Cheminert™ Model C2 Microbore Injectors as injectors or switching valves. It can be actuated manually or with a microelectronic actuator. With 1/16 in. fittings and 0.010 in. ports, it is available in six-port or ten-port configurations and includes a 5μL sample loop.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) Sample 10 μL
Type Sample Injector Loop
Stator Material Stainless Steel